Assay Details
| Gene Symbol: BMS1P20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DB472458.1 | - | 2 | 323 | 164 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390289.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000390290.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000413293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000426066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | ENST00000436538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009206.hg.1 | NR_027293.2 | Refseq |

BMS1P20
Genomic Map