Assay Details
| Gene Symbol: RAB44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257357.1 | NP_001244286.1 | 11 - 12 | 2875 | 72 | |
| XM_011514600.2 | XP_011512902.1 | 11 - 12 | 2972 | 72 | ||
| XM_011514601.2 | XP_011512903.1 | 12 - 13 | 3019 | 72 | ||
| XM_011514602.2 | XP_011512904.1 | 11 - 12 | 2939 | 72 | ||
| XM_017010855.1 | XP_016866344.1 | 11 - 12 | 4411 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154475.hg.1 | TC0600007848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000229824.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | ENST00000612677.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007848.hg.1 | NM_001257357.1 | Refseq |

RAB44
Genomic Map