Assay Details
| Gene Symbol: OCSTAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080721.2 | NP_542452.1 | 2 - 3 | 1065 | 68 | |
| XM_011528532.2 | XP_011526834.1 | 1 - 2 | 875 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000904.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 99398 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 141751 | AK310440.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 141751 | BC004262.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 141751 | NM_001080543.1 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 226670 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 225110 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3403942 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3010 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3009 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 32922 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P21145239 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3403942 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3403942 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839858 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839874 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839859 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839858 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17195767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839874 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839865 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191774 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191768 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191771 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191791 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11191766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31839859 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191744 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191775 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191761 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195766 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191793 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191765 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191743 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191779 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191794 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191781 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191740 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191768 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191788 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191786 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191791 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17195762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11191769 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P21145239 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03525403 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8066755 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8066756 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8066754 | - | - |
| Affymetrix Human Genome U219 Array | 11740498_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11740498_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012433.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012433.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012433.hg.1 | TC20000904.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012432.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012432.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012432.hg.1 | TC20000904.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012434.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012434.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012434.hg.1 | TC20000904.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000165023.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165023.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165023.hg.1 | TC2000009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000165023.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165023.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165024.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165024.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165024.hg.1 | TC2000009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000165024.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165024.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165021.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165021.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165021.hg.1 | TC2000009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000165021.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165021.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009275.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165022.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165022.hg.1 | NM_080721.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000165022.hg.1 | TC2000009275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000165022.hg.1 | ENST00000279028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000165022.hg.1 | NM_080721.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017747 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 211903 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1677441 | NM_080721.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677441 | NM_080721.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677441 | NM_080721.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1677441 | NM_080721.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1677441 | NM_080721.1 | human_transcriptome |

OCSTAMP
Genomic Map