| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050775.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050775.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050775.hg.1 |
TC01003292.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050776.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050776.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050776.hg.1 |
TC01003292.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
ENST00000555188.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018495.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243227.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243227.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243227.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243227.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243227.hg.1 |
TC0100018495.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100243228.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243228.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243228.hg.1 |
ENST00000304760.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243228.hg.1 |
NM_001010846.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243228.hg.1 |
TC0100018495.hg.1 |
Clariom D TC |