Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012560.hg.1 |
TC05000863.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012560.hg.1 |
ENST00000420481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012560.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011510.hg.1 |
ENST00000420261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011510.hg.1 |
ENST00000420261.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011510.hg.1 |
ENST00000429032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011510.hg.1 |
ENST00000420261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011510.hg.1 |
ENST00000420261.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011510.hg.1 |
ENST00000429032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164401.hg.1 |
ENST00000522232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164401.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164401.hg.1 |
ENST00000522232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164401.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164401.hg.1 |
TC0500009201.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500164402.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164402.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164402.hg.1 |
TC0500009201.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500164400.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164400.hg.1 |
NR_038443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164400.hg.1 |
TC0500009201.hg.1 |
Clariom D TC |