Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052909.3 | NP_443141.3 | 1 - 2 | 460 | 105 | |
| XM_006714446.3 | XP_006714509.1 | 3 - 4 | 1598 | 105 | ||
| XM_011513968.2 | XP_011512270.1 | 3 - 4 | 1598 | 105 | ||
| XM_011513969.2 | XP_011512271.1 | 3 - 4 | 1475 | 105 | ||
| XM_011513970.1 | XP_011512272.1 | 3 - 4 | 1187 | 105 | ||
| XM_011513971.2 | XP_011512273.1 | 3 - 4 | 1543 | 105 | ||
| XM_011513972.2 | XP_011512274.1 | 3 - 4 | 1599 | 105 | ||
| XM_011513973.1 | XP_011512275.1 | 3 - 4 | 1596 | 105 | ||
| XM_017009112.1 | XP_016864601.1 | 3 - 4 | 1599 | 105 | ||
| GenBank mRNA | AB067496.2 | - | 1 - 2 | 460 | 105 | |
| BC008352.1 | - | 1 - 2 | 152 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000002.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05000004.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000004.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000004.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000005.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000005.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000005.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000006.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000006.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000006.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000007.hg.1 | TC05000002.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000007.hg.1 | ENST00000283426.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000007.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144062.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144062.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144062.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144062.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144062.hg.1 | TC0500006435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500144064.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144064.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144064.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144064.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144064.hg.1 | TC0500006435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144065.hg.1 | TC0500006435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | ENST00000504041.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006435.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | ENST00000283426.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | ENST00000502646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | NM_052909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500144066.hg.1 | TC0500006435.hg.1 | Clariom D TC |

PLEKHG4B
Genomic Map