| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000011.hg.1 |
TC05000002.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000011.hg.1 |
ENST00000283426.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05000011.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500144070.hg.1 |
TC0500006435.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000283426.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000502646.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
ENST00000504041.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006435.hg.1 |
NM_052909.4 |
Refseq |