Assay Details
| Gene Symbol: STK17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004226.3 | NP_004217.1 | 1 - 2 | 274 | 66 | |
| GenBank mRNA | AB011421.1 | - | 1 - 2 | 211 | 66 | |
| BC016040.1 | - | 1 - 2 | 213 | 4575012 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002637.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB011421.1,BC016040.1,NM_004226.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042258.hg.1 | TC02002637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042258.hg.1 | ENST00000263955.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042258.hg.1 | ENST00000409228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042258.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042264.hg.1 | TC02002637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042264.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042256.hg.1 | TC02002637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042256.hg.1 | ENST00000263955.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042256.hg.1 | ENST00000409228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042256.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042257.hg.1 | TC02002637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042257.hg.1 | ENST00000263955.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042257.hg.1 | ENST00000409228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042257.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042262.hg.1 | TC02002637.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042262.hg.1 | ENST00000263955.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02042262.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212088.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212088.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212088.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212088.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212088.hg.1 | TC0200015314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212082.hg.1 | TC0200015314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212084.hg.1 | TC0200015314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212080.hg.1 | TC0200015314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200212090.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212090.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200212090.hg.1 | TC0200015314.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000263955.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000409228.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000420683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000449152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | ENST00000606789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015314.hg.1 | NM_004226.3 | Refseq |

STK17B
Genomic Map