Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000488.3 | NP_000479.1 | 2 - 3 | 524 | 170 | |
| XM_005245198.3 | XP_005245255.1 | 3 - 4 | 690 | 170 | ||
| GenBank mRNA | AF130100.1 | - | 4 - 5 | 638 | 170 | |
| AK312654.1 | - | 2 - 3 | 473 | 170 | ||
| BC022309.1 | - | 2 - 3 | 504 | 4734914 | 170 | |
| D29832.1 | - | 2 - 3 | 429 | 170 | ||
| KP780080.1 | - | 2 - 3 | 405 | 170 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003535.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01054895.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054895.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054895.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054895.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054895.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054894.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054894.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054894.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054894.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054894.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054897.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054897.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054897.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054897.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054897.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054892.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054892.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054892.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054892.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054893.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054893.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054893.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054893.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054893.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054898.hg.1 | TC01003535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054898.hg.1 | ENST00000351522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054898.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01054898.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225092.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225094.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225091.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225089.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225096.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225090.hg.1 | TC0100016445.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000487183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016445.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000367698.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000494024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | ENST00000617423.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | NM_000488.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225095.hg.1 | TC0100016445.hg.1 | Clariom D TC |

SERPINC1
Genomic Map