| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020840.hg.1 |
TC08001592.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020840.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020840.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020839.hg.1 |
TC08001592.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020839.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020839.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179370.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179370.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179370.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179370.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179370.hg.1 |
TC0800011690.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800179371.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179371.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179371.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179371.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179371.hg.1 |
TC0800011690.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
ENST00000325995.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011690.hg.1 |
NM_001081675.2 |
Refseq |