Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016988.hg.1 |
TC06001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016988.hg.1 |
ENST00000376442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016988.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016988.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002399.hg.1 |
TC6_qbl_hap6000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002399.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002399.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001024.hg.1 |
TC6_apd_hap1000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001024.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1001024.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001946.hg.1 |
TC6_mann_hap4000116.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001946.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001946.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002116.hg.1 |
TC6_mcf_hap5000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002116.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002116.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002426.hg.1 |
TC6_cox_hap2000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002426.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002426.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002356.hg.1 |
TC6_dbb_hap3000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002356.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002356.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002200.hg.1 |
TC6_ssto_hap7000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002200.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002200.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176680.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176680.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176680.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176680.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176680.hg.1 |
TC0600011375.hg.1 |
Clariom D TC |