| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041780.hg.1 |
TC02002612.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041780.hg.1 |
ENST00000374866.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041780.hg.1 |
ENST00000452536.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02041780.hg.1 |
NM_000393.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
ENST00000374866.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
ENST00000618828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
NM_000393.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
ENST00000374866.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
ENST00000618828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
NM_000393.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200211330.hg.1 |
TC0200015204.hg.1 |
Clariom D TC |