Assay Details
| Gene Symbol: COL5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000393.3 | NP_000384.2 | 41 - 42 | 3044 | 105 | |
| XM_011510573.2 | XP_011508875.1 | 41 - 42 | 2826 | 105 | ||
| GenBank mRNA | AB209045.1 | - | 41 - 42 | 2906 | 105 | |
| BC043613.1 | - | 3 - 4 | 139 | 105 | ||
| BC086874.1 | - | 25 - 26 | 2033 | 30708812 | 105 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002612.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC043613.1 transcripts.
Set Membership: |
|

COL5A2
Genomic Map