Assay Details
| Gene Symbol: MUC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002457.3 | NP_002448.3 | 15223 | 71 | ||
| GenBank mRNA | L21998.1 | - | 15223 | 71 | ||
| M94132.1 | - | 18 - 19 | 2611 | 71 | ||
More Information
Set Membership: |
|

MUC2
Genomic Map