Assay Details
| Gene Symbol: CLIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287527.1 | NP_001274456.1 | 4 - 5 | 617 | 156 | |
| NM_001287528.1 | NP_001274457.1 | 4 - 5 | 653 | 156 | ||
| NM_024692.5 | NP_078968.3 | 4 - 5 | 621 | 156 | ||
| XM_005264562.2 | XP_005264619.1 | 4 - 5 | 648 | 156 | ||
| XM_005264563.1 | XP_005264620.1 | 4 - 5 | 653 | 156 | ||
| XM_006712104.2 | XP_006712167.1 | 4 - 5 | 693 | 156 | ||
| XM_006712105.3 | XP_006712168.1 | 4 - 5 | 535 | 156 | ||
| XM_011533109.1 | XP_011531411.1 | 4 - 5 | 401 | 156 | ||
| XM_011533110.1 | XP_011531412.1 | 3 - 4 | 454 | 156 | ||
| XM_017004958.1 | XP_016860447.1 | 4 - 5 | 480 | 156 | ||
| XM_017004959.1 | XP_016860448.1 | 4 - 5 | 535 | 156 | ||
| GenBank mRNA | AB209042.1 | - | 4 - 5 | 2232 | 156 | |
| AF433661.1 | - | 4 - 5 | 629 | 156 | ||
| AK291595.1 | - | 4 - 5 | 698 | 156 | ||
| AK302325.1 | - | 4 - 5 | 603 | 156 | ||
| AK304384.1 | - | 3 - 4 | 447 | 156 | ||
| AL834443.1 | - | 4 - 5 | 595 | 156 | ||
| BC013862.1 | - | 4 - 5 | 601 | 156 | ||
| BC015310.1 | - | 2 - 3 | 227 | 156 | ||
| BC126167.1 | - | 3 - 4 | 391 | 8991883 | 156 | |
| BC136331.1 | - | 3 - 4 | 391 | 9020318 | 156 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_011533110.1 transcripts.
Set Membership: |
|

CLIP4
Genomic Map