Assay Details
| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017437.3 | NP_001017437.2 | 2 - 3 | 653 | 82 | |
| NM_001318335.1 | NP_001305264.1 | 2 - 3 | 653 | 82 | ||
| XM_006724278.3 | XP_006724341.1 | 2 - 3 | 260 | 82 | ||
| XM_006724279.3 | XP_006724342.1 | 2 - 3 | 260 | 82 | ||
| XM_006724280.3 | XP_006724343.1 | 2 - 3 | 260 | 82 | ||
| XM_011530263.2 | XP_011528565.1 | 2 - 3 | 313 | 82 | ||
| XM_011530266.2 | XP_011528568.1 | 2 - 3 | 260 | 82 | ||
| XM_011530267.2 | XP_011528569.1 | 2 - 3 | 260 | 82 | ||
| XM_011530268.2 | XP_011528570.1 | 2 - 3 | 260 | 82 | ||
| XM_011530270.2 | XP_011528572.1 | 2 - 3 | 260 | 82 | ||
| GenBank mRNA | BC018040.1 | - | 2 - 3 | 653 | 82 | |
| DB636414.1 | - | 2 - 3 | 281 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000208.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001017437.3,NM_001318335.1,XM_006724278.3,XM_006724279.3,XM_006724280.3,XM_011530263.2,XM_011530266.2,XM_011530267.2,XM_011530268.2,XM_011530270.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003894.hg.1 | ENST00000338306.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003894.hg.1 | ENST00000399824.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003894.hg.1 | ENST00000405659.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003894.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003894.hg.1 | TC22000208.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000475975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | ENST00000482413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007051.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | TC2200007051.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | NM_001017437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000338306.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000399824.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000405659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000430839.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | ENST00000445005.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200149300.hg.1 | NM_001017437.3 | Refseq |

CCDC157
Genomic Map