Assay Details
| Gene Symbol: GBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004120.4 | NP_004111.2 | 9 - 10 | 1775 | 90 | |
| GenBank mRNA | AK226018.1 | - | 9 - 10 | 1701 | 90 | |
| AK314325.1 | - | 9 - 10 | 1730 | 90 | ||
| BC073163.1 | - | 9 - 10 | 1712 | 6151056 | 90 | |
| CR457062.1 | - | 8 - 9 | 1461 | 90 | ||
| M55543.1 | - | 9 - 10 | 1617 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006361.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01044400.hg.1 | TC01006361.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044400.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044400.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044403.hg.1 | TC01006361.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044403.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044403.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044403.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044404.hg.1 | TC01006361.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044404.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044404.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044404.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242305.hg.1 | TC0100018451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242308.hg.1 | TC0100018451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242309.hg.1 | TC0100018451.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000370466.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000463660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | ENST00000493802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018451.hg.1 | NM_004120.4 | Refseq |

GBP2
Genomic Map