Assay Details
Gene Symbol: IFT57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018010.3 | NP_060480.1 | 4 - 5 | 832 | 118 | |
GenBank mRNA | AB014762.1 | - | 4 - 5 | 584 | 118 | |
AF139576.1 | - | 4 - 5 | 662 | 118 | ||
AF245220.1 | - | 4 - 5 | 726 | 118 | ||
AK001009.1 | - | 4 - 5 | 671 | 118 | ||
AK298854.1 | - | 4 - 5 | 730 | 118 | ||
BC011899.2 | - | 4 - 5 | 681 | 4053749 | 118 | |
CR457251.1 | - | 4 - 5 | 584 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
