Assay Details
| Gene Symbol: STPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006711025.2 | XP_006711088.1 | 2 - 3 | 203 | 114 | |
| XM_017002768.1 | XP_016858257.1 | 3 - 4 | 431 | 114 | ||
| XM_017002770.1 | XP_016858259.1 | 3 - 4 | 431 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006711025.2, XM_017002768.1, XM_017002770.1 transcripts.
Set Membership: |
|

STPG1
Genomic Map