Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011925.hg.1 |
ENST00000306117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011925.hg.1 |
ENST00000537075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011925.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011925.hg.1 |
TC20000873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011927.hg.1 |
ENST00000306117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011927.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011927.hg.1 |
TC20000873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011928.hg.1 |
ENST00000306117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011928.hg.1 |
ENST00000537075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011928.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011928.hg.1 |
TC20000873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011929.hg.1 |
ENST00000306117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011929.hg.1 |
ENST00000537075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011929.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011929.hg.1 |
TC20000873.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011924.hg.1 |
ENST00000306117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011924.hg.1 |
ENST00000537075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011924.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20011924.hg.1 |
TC20000873.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
TC2000009209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164417.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164421.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164421.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164421.hg.1 |
TC2000009209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164421.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164421.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
TC2000009209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164422.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164423.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164423.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164423.hg.1 |
TC2000009209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164423.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164423.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009209.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
NM_002251.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
TC2000009209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
ENST00000306117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
ENST00000537075.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164418.hg.1 |
NM_002251.4 |
Refseq |