Assay Details
| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528480.2 | XP_011526782.1 | 2 - 3 | 313 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528480.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acyl-CoA metabolic process
bile acid biosynthetic process
viral process
peroxisome fission
fatty acid beta-oxidation using acyl-CoA oxidase
long-chain fatty-acyl-CoA biosynthetic process
alpha-linolenic acid metabolic process
dicarboxylic acid catabolic process
negative regulation of CD4 biosynthetic process
acetyl-CoA hydrolase activity
receptor binding
protein binding
CoA hydrolase activity
palmitoyl-CoA hydrolase activity
choloyl-CoA hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
long-chain acyl-CoA hydrolase activity
acyl-CoA metabolic process
bile acid biosynthetic process
viral process
peroxisome fission
fatty acid beta-oxidation using acyl-CoA oxidase
long-chain fatty-acyl-CoA biosynthetic process
alpha-linolenic acid metabolic process
dicarboxylic acid catabolic process
negative regulation of CD4 biosynthetic process
acetyl-CoA hydrolase activity
receptor binding
protein binding
CoA hydrolase activity
palmitoyl-CoA hydrolase activity
choloyl-CoA hydrolase activity
acyl-CoA hydrolase activity
carboxylic ester hydrolase activity
medium-chain acyl-CoA hydrolase activity
long-chain acyl-CoA hydrolase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000217455.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000426915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000457981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000461272.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000481938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000483141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484783.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000484975.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000486165.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000487205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000488679.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | ENST00000493118.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009245.hg.1 | NM_005469.3 | Refseq |

ACOT8
Genomic Map