Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024641.hg.1 |
ENST00000392676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024641.hg.1 |
ENST00000392677.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024641.hg.1 |
NM_018672.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024641.hg.1 |
NM_172232.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17024641.hg.1 |
TC17001830.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000392676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000586811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000586995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000588106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000588877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000591234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
NM_018672.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
NM_172232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000392676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000586811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000586995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000588106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000588877.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
ENST00000591234.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
NM_018672.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
NM_172232.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700207543.hg.1 |
TC1700012460.hg.1 |
Clariom D TC |