Assay Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015543.1 | NP_056358.1 | 3 - 4 | 571 | 135 | |
| GenBank mRNA | AL050268.1 | - | 3 - 4 | 375 | 135 | |
| CR533479.1 | - | 3 - 4 | 185 | 135 | ||
More Information
Set Membership: |
|

RAB1A
Genomic Map