Assay Details
Gene Symbol: LHCGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000233.3 | NP_000224.2 | 2 - 3 | 308 | 87 | |
GenBank mRNA | M63108.1 | - | 2 - 3 | 246 | 87 | |
S57793.1 | - | 2 - 3 | 238 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001830.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
phospholipase C-activating G-protein coupled receptor signaling pathway
male gonad development
hormone-mediated signaling pathway
ovulation cycle process
male genitalia development
positive regulation of inositol trisphosphate biosynthetic process
luteinizing hormone signaling pathway
positive regulation of cAMP-mediated signaling
cognition
cellular response to gonadotropin stimulus
luteinizing hormone receptor activity
G-protein coupled peptide receptor activity
choriogonadotropin hormone receptor activity
choriogonadotropin hormone binding
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
phospholipase C-activating G-protein coupled receptor signaling pathway
male gonad development
hormone-mediated signaling pathway
ovulation cycle process
male genitalia development
positive regulation of inositol trisphosphate biosynthetic process
luteinizing hormone signaling pathway
positive regulation of cAMP-mediated signaling
cognition
cellular response to gonadotropin stimulus
luteinizing hormone receptor activity
G-protein coupled peptide receptor activity
choriogonadotropin hormone receptor activity
choriogonadotropin hormone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | TC02001830.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | ENST00000294954.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | ENST00000344775.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | ENST00000401907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | ENST00000403273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | ENST00000405626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02029293.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | ENST00000477576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016666.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000294954.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000401907.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000403273.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000405626.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | ENST00000428232.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | NM_000233.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200226279.hg.1 | TC0200016666.hg.1 | Clariom D TC |
