Assay Details
| Gene Symbol: DCTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135040.2 | NP_001128512.1 | 23 - 24 | 3454 | 77 | |
| NM_001135041.2 | NP_001128513.1 | 21 - 22 | 3177 | 77 | ||
| NM_001190836.1 | NP_001177765.1 | 23 - 24 | 3397 | 77 | ||
| NR_033935.1 | 22 - 23 | 3480 | 77 | |||
| GenBank mRNA | AB209625.1 | - | 15 - 16 | 4449 | 77 | |
| AK297286.1 | - | 23 - 24 | 3149 | 77 | ||
| AK314352.1 | - | 23 - 24 | 3398 | 77 | ||
| AK315014.1 | - | 21 - 22 | 2969 | 77 | ||
| BC071583.1 | - | 21 - 22 | 3094 | 4817849 | 77 | |
| BX647925.1 | - | 22 - 23 | 3241 | 77 | ||
| BX647972.1 | - | 22 - 23 | 3260 | 77 | ||
More Information
Set Membership: |
|

DCTN1
Genomic Map