Assay Details
| Gene Symbol: TAF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003487.3 | NP_003478.1 | 4 - 5 | 286 | 93 | |
| GenBank mRNA | AK313223.1 | - | 4 - 5 | 252 | 93 | |
| X98893.1 | - | 4 - 5 | 217 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

TAF15
Genomic Map