Assay Details
| Gene Symbol: SUCLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177599.1 | NP_001171070.1 | 1 - 2 | 107 | 60 | |
| NM_003848.3 | NP_003839.2 | 1 - 2 | 107 | 60 | ||
| XM_017007415.1 | XP_016862904.1 | 1 - 2 | 103 | 60 | ||
| XM_017007416.1 | XP_016862905.1 | 1 - 2 | 103 | 60 | ||
| XM_017007417.1 | XP_016862906.1 | 1 - 2 | 103 | 60 | ||
| XM_017007418.1 | XP_016862907.1 | 1 - 2 | 103 | 60 | ||
| XM_017007419.1 | XP_016862908.1 | 1 - 2 | 103 | 60 | ||
| XM_017007420.1 | XP_016862909.1 | 1 - 2 | 103 | 60 | ||
| GenBank mRNA | AK294598.1 | - | 1 - 2 | 100 | 60 | |
| AK310527.1 | - | 1 - 2 | 103 | 60 | ||
| BC007716.2 | - | 1 - 2 | 70 | 4303307 | 60 | |
| BC068602.1 | - | 1 - 2 | 108 | 5274341 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001533.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC007716.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | TC03001533.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | ENST00000307227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | ENST00000493112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | ENST00000541608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027942.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008494.hg.1 | ENST00000307241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | TC03001533.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | ENST00000307227.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | ENST00000493112.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | ENST00000541608.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03027941.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007017.hg.1 | ENST00000514739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007017.hg.1 | ENST00000514739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185718.hg.1 | TC0300011436.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000460567.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011436.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008494.hg.1 | ENST00000307241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007017.hg.1 | ENST00000514739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007017.hg.1 | ENST00000514739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000307227.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000492795.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | ENST00000493112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | NM_001177599.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | NM_003848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300185719.hg.1 | TC0300011436.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 192798 | - | - |

SUCLG2
Genomic Map