Assay Details
| Gene Symbol: RANBP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161429.2 | NP_001154901.1 | 11 - 12 | 1476 | 125 | |
| NM_001323273.1 | NP_001310202.1 | 10 - 11 | 1394 | 125 | ||
| NM_001323274.1 | NP_001310203.1 | 11 - 12 | 1469 | 125 | ||
| NM_001323275.1 | NP_001310204.1 | 11 - 12 | 1409 | 125 | ||
| NM_001323276.1 | NP_001310205.1 | 11 - 12 | 1409 | 125 | ||
| NM_001323277.1 | NP_001310206.1 | 10 - 11 | 1394 | 125 | ||
| NM_001323278.1 | NP_001310207.1 | 10 - 11 | 1334 | 125 | ||
| NM_001323279.1 | NP_001310208.1 | 9 - 10 | 1322 | 125 | ||
| NM_001323280.1 | NP_001310209.1 | 10 - 11 | 1334 | 125 | ||
| NM_145000.4 | NP_659437.3 | 10 - 11 | 1401 | 125 | ||
| XM_006714452.3 | XP_006714515.1 | 11 - 12 | 1503 | 125 | ||
| XM_006714453.3 | XP_006714516.1 | 11 - 12 | 1443 | 125 | ||
| XM_006714455.3 | XP_006714518.1 | 10 - 11 | 1368 | 125 | ||
| GenBank mRNA | AK302947.1 | - | 11 - 12 | 1467 | 125 | |
| BC047660.1 | - | 10 - 11 | 1394 | 5261736 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001271.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017805.hg.1 | TC05001271.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017805.hg.1 | ENST00000296604.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017805.hg.1 | ENST00000502994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017805.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017805.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500173112.hg.1 | TC0500010498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |

RANBP3L
Genomic Map