Assay Details
| Gene Symbol: UNC80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005246476.1 | XP_005246533.1 | 27 - 28 | 4563 | 84 | |
| XM_011511004.1 | XP_011509306.1 | 27 - 28 | 4605 | 84 | ||
| XM_011511005.1 | XP_011509307.1 | 27 - 28 | 4605 | 84 | ||
| XM_011511006.1 | XP_011509308.1 | 27 - 28 | 4599 | 84 | ||
| XM_011511007.1 | XP_011509309.1 | 27 - 28 | 4569 | 84 | ||
| XM_011511008.1 | XP_011509310.1 | 27 - 28 | 4605 | 84 | ||
| XM_011511010.2 | XP_011509312.1 | 27 - 28 | 4605 | 84 | ||
| XM_011511012.1 | XP_011509314.1 | 27 - 28 | 4605 | 84 | ||
| XM_017003884.1 | XP_016859373.1 | 27 - 28 | 4590 | 84 | ||
| XM_017003885.1 | XP_016859374.1 | 27 - 28 | 4563 | 84 | ||
| XM_017003886.1 | XP_016859375.1 | 27 - 28 | 4605 | 84 | ||
| XM_017003891.1 | XP_016859380.1 | 26 - 27 | 4624 | 84 | ||
| XM_017003892.1 | XP_016859381.1 | 14 - 15 | 3408 | 84 | ||
| XM_017003893.1 | XP_016859382.1 | 27 - 28 | 4605 | 84 | ||
| XM_017003894.1 | XP_016859383.1 | 5 - 6 | 836 | 84 | ||
More Information
Set Membership: |
|

UNC80
Genomic Map