Assay Details
| Gene Symbol: DHX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033905.1 | 4 - 5 | 376 | 61 | ||
| GenBank mRNA | AK296506.1 | - | 4 - 5 | 354 | 61 | |
| AK296793.1 | - | 4 - 5 | 346 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296793.1 transcripts.
Set Membership: |
|

DHX35
Genomic Map