Assay Details
| Gene Symbol: CPNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152927.2 | NP_690904.1 | 2 - 3 | 283 | 84 | |
| NM_152928.2 | NP_690905.1 | 2 - 3 | 251 | 84 | ||
| GenBank mRNA | BG701713.1 | - | 2 - 3 | 233 | 84 | |
| BI917584.1 | - | 2 - 3 | 214 | 84 | ||
| DA983782.1 | - | 2 - 3 | 195 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_152927.2, NM_152928.2 transcripts.
Set Membership: |
|

CPNE1
Genomic Map