Assay Details
| Gene Symbol: LOC100147773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC009228.2 | - | 1 | 726 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane and coiled-coil domains 1 | TMCO1 |
HP10122, PCIA3, PNAS-136, TMCC4 |
-460.0 | Sense | Chr. 1 - 165724291 - 165768922 on NCBI Build GRCh38 |
| Downstream | uridine-cytidine kinase 2 | UCK2 |
TSA903, UK, UMPK |
57.6kb | Anti-sense | Chr. 1 - 165827495 - 165911618 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100147773
Genomic Map