Assay Details
| Gene Symbol: PTPRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_052021.1 | 3 | 1035 | 83 | ||
| GenBank mRNA | BC017863.1 | - | 3 | 907 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PTPRC
Genomic Map