Assay Details
| Gene Symbol: NR5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509381.2 | XP_011507683.1 | 1 - 2 | 1763 | 106 | |
| GenBank mRNA | AF124248.1 | - | 2 - 3 | 102 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC400800 | LOC400800 | -643.7kb | Sense | Chr. 1 - 199148598 - 199393307 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 862 | LINC00862 |
C1orf98, SMIM16 |
302.7kb | Anti-sense | Chr. 1 - 200342544 - 200373792 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011509381.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
bile acid metabolic process
hormone-mediated signaling pathway
embryo development
tissue development
intracellular receptor signaling pathway
epithelial cell differentiation
regulation of cell proliferation
homeostatic process
cholesterol homeostasis
steroid hormone mediated signaling pathway
positive regulation of viral genome replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
pancreas morphogenesis
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
phospholipid binding
zinc ion binding
sequence-specific DNA binding
transcription regulatory region DNA binding
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
bile acid metabolic process
hormone-mediated signaling pathway
embryo development
tissue development
intracellular receptor signaling pathway
epithelial cell differentiation
regulation of cell proliferation
homeostatic process
cholesterol homeostasis
steroid hormone mediated signaling pathway
positive regulation of viral genome replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
pancreas morphogenesis
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
phospholipid binding
zinc ion binding
sequence-specific DNA binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000236914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367357.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000367362.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000447034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000474307.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | ENST00000544748.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_001276464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_003822.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011129.hg.1 | NM_205860.2 | Refseq |

NR5A2
Genomic Map