| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178240.hg.1 |
TC0100010625.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010625.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
ENST00000340030.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
ENST00000367721.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
NM_000639.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
NM_001302746.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100178239.hg.1 |
TC0100010625.hg.1 |
Clariom D TC |