Assay Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012278.2 | NP_036410.1 | 1 - 2 | 124 | 62 | |
| XM_017029405.1 | XP_016884894.1 | 1 - 2 | 597 | 62 | ||
| GenBank mRNA | AF110225.1 | - | 1 - 2 | 78 | 62 | |
| AF140690.1 | - | 1 - 2 | 94 | 62 | ||
| DA895605.1 | - | 1 - 2 | 127 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000386.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017029405.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005178.hg.1 | TC0X000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005178.hg.1 | ENST00000373829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005178.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005178.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005181.hg.1 | TC0X000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005181.hg.1 | ENST00000373829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005181.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005181.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005180.hg.1 | TC0X000386.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005180.hg.1 | ENST00000373829.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005180.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X005180.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151988.hg.1 | TC0X00007562.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151986.hg.1 | TC0X00007562.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00151990.hg.1 | TC0X00007562.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000373829.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000465388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000475413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000483897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | ENST00000538820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_001303277.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007562.hg.1 | NM_012278.3 | Refseq |

ITGB1BP2
Genomic Map