Assay Details
| Gene Symbol: SLAMF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003037.3 | NP_003028.1 | 1 - 2 | 443 | 97 | |
| NR_104399.1 | 1 - 2 | 443 | 97 | |||
| NR_104400.1 | 1 - 2 | 443 | 97 | |||
| NR_104401.1 | 1 - 2 | 443 | 97 | |||
| XM_005245456.3 | XP_005245513.1 | 1 - 2 | 669 | 97 | ||
| XM_011509905.2 | XP_011508207.1 | 1 - 2 | 669 | 97 | ||
| XM_017002129.1 | XP_016857618.1 | 1 - 2 | 669 | 97 | ||
| XM_017002130.1 | XP_016857619.1 | 1 - 2 | 669 | 97 | ||
| XM_017002131.1 | XP_016857620.1 | 1 - 2 | 669 | 97 | ||
| GenBank mRNA | AK304237.1 | - | 1 - 2 | 153 | 97 | |
| AK314244.1 | - | 1 - 2 | 387 | 97 | ||
| AY040554.1 | - | 1 - 2 | 77 | 97 | ||
| BC012602.1 | - | 1 - 2 | 430 | 97 | ||
| BC132792.1 | - | 1 - 2 | 109 | 40146814 | 97 | |
| BC144498.1 | - | 1 - 2 | 109 | 97 | ||
| DB133023.1 | - | 1 - 2 | 423 | 97 | ||
| KF471075.1 | - | 1 - 2 | 90 | 97 | ||
| U33017.1 | - | 1 - 2 | 210 | 97 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_011509905.2, XM_017002131.1 transcripts.
This assay is in the 5'UTR region of the XM_011509905.2, XM_017002131.1 transcripts.
Set Membership: |
|

SLAMF1
Genomic Map