Assay Details
Gene Symbol: EPB41L4A-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027706.1 | 1 | 1219 | 103 | ||
GenBank mRNA | AK002097.1 | - | 1 | 1219 | 103 | |
BX089466.1 | - | 1 | 465 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000530.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04196162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24375354 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24375352 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04196158 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24375349 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24375353 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 123742 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 2793 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P69941 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P188975 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P69941 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 8900 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 15473 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P112696 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P112696 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 19694 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P69941 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00800270 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P69941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196124 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196141 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375340 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196125 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196149 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375328 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375335 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196124 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375328 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196141 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196129 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375340 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196125 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196149 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375335 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04196134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375325 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24375324 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P69941 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P188975 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P112696 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P112696 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P112696 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 68612 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P112696 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 4915245 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 4915245 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196158 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196132 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196123 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196129 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196140 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196149 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196144 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196124 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196156 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196151 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15731308 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196138 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196162 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196153 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15731306 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15731303 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196126 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196145 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196142 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196160 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196134 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196155 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04196147 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P112696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8107329 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8107328 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220364_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11745308_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11745308_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR05007351.hg.1 | TC05000530.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05007351.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008324.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008324.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008324.hg.1 | ENST00000624967.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008324.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008324.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156966.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156966.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156966.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500156966.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500156966.hg.1 | TC0500008324.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500008324.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008324.hg.1 | ENST00000623705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008324.hg.1 | ENST00000624967.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008324.hg.1 | NR_027706.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008324.hg.1 | NR_027706.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 220364_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 220364_at | - | - |
Human Genome U133A 2.0 Array | 220364_PM_at | - | - |
Human Genome U133A 2.0 Array | 220364_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw536984 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220364_at1 | - | - |
Human Genome U95 Set | 56767_PM_at | - | - |
Human Genome U95 Set | 56767_at | - | - |
Human Genome U133 Set | 220364_at | - | - |
Human Genome U133 Set | 220364_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028622 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A028622 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 174371 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1689831 | BX089466.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1689831 | NR_027706.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1689831 | AK002097.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689831 | BX089466.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689831 | NR_027706.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689831 | AK002097.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1689831 | BX089466.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1689831 | NR_027706.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1689831 | AK002097.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689831 | BX089466.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689831 | NR_027706.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689831 | AK002097.1 | human_transcriptome |
