Assay Details
Gene Symbol: C1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300834.1 | NP_001287763.1 | 1 - 2 | 166 | 70 | |
NM_001300835.1 | NP_001287764.1 | 1 - 2 | 166 | 70 | ||
NM_174896.3 | NP_777556.1 | 1 - 2 | 166 | 70 | ||
XM_011540712.2 | XP_011539014.1 | 2 - 3 | 5646 | 70 | ||
XM_017000323.1 | XP_016855812.1 | 2 - 3 | 1515 | 70 | ||
XM_017000324.1 | XP_016855813.1 | 2 - 3 | 5646 | 70 | ||
GenBank mRNA | BC017973.1 | - | 1 - 2 | 53 | 4693393 | 70 |
BG545926.1 | - | 1 - 2 | 56 | 70 | ||
HX785231.1 | - | 1 - 2 | 48 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000978.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC017973.1,NM_001300834.1,NM_001300835.1,NM_174896.3,XM_011540712.2,XM_017000323.1,XM_017000324.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015606.hg.1 | TC01000978.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015606.hg.1 | ENST00000343534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015606.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015606.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015609.hg.1 | TC01000978.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015609.hg.1 | ENST00000343534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015609.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015609.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015609.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015608.hg.1 | TC01000978.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015608.hg.1 | ENST00000343534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015608.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015608.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015608.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01015611.hg.1 | TC01000978.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01015611.hg.1 | ENST00000343534.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015611.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01015611.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167783.hg.1 | TC0100009417.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167786.hg.1 | TC0100009417.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | ENST00000343534.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | ENST00000369718.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_001300834.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_001300835.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | NM_174896.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100167782.hg.1 | TC0100009417.hg.1 | Clariom D TC |
