Assay Details
| Gene Symbol: PHYHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099335.1 | NP_001092805.1 | 3 - 4 | 864 | 96 | |
| NM_014759.3 | NP_055574.3 | 2 - 3 | 742 | 96 | ||
| XM_006716416.1 | XP_006716479.1 | 2 - 3 | 537 | 96 | ||
| GenBank mRNA | AK098743.1 | - | 3 - 4 | 487 | 96 | |
| BC034034.1 | - | 2 - 3 | 495 | 4793002 | 96 | |
| BC035940.1 | - | 2 - 3 | 495 | 4793228 | 96 | |
| D87463.1 | - | 3 - 4 | 573 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001036.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | TC08001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | ENST00000321613.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | ENST00000454243.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | ENST00000456132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013238.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013235.hg.1 | TC08001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013235.hg.1 | ENST00000321613.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013235.hg.1 | ENST00000454243.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013235.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013235.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166810.hg.1 | TC0800009833.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166812.hg.1 | TC0800009833.hg.1 | Clariom D TC |

PHYHIP
Genomic Map