| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000257336.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000448849.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000474443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000474443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000474443.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000481069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000481069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000481069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000481069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000481069.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000490317.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
ENST00000491929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_001159596.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009993.hg.1 |
NM_017693.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009992.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166929.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166929.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166929.hg.1 |
ENST00000602836.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166929.hg.1 |
NM_001204425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166929.hg.1 |
TC1300009992.hg.1 |
Clariom D TC |