Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
ENST00000333229.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
ENST00000342449.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
ENST00000380783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
ENST00000380800.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
NM_018963.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005370.hg.1 |
TC21000459.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000333229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000342449.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000380800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000424441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000446924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000473813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
NM_018963.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
NM_033656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
TC2100008165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000333229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000342449.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000380800.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000424441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000446924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
ENST00000473813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
NM_018963.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100155074.hg.1 |
NM_033656.3 |
Refseq |