Assay Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007246.2 | NP_001007247.1 | 3 - 4 | 430 | 108 | |
NM_018963.4 | NP_061836.2 | 3 - 4 | 430 | 108 | ||
NM_033656.3 | NP_387505.1 | 3 - 4 | 430 | 108 | ||
XM_011529611.1 | XP_011527913.1 | 3 - 4 | 463 | 108 | ||
XM_017028373.1 | XP_016883862.1 | 4 - 5 | 235 | 108 | ||
XM_017028375.1 | XP_016883864.1 | 3 - 4 | 463 | 108 | ||
GenBank mRNA | AB080586.1 | - | 3 - 4 | 234 | 108 | |
AB080587.1 | - | 3 - 4 | 234 | 108 | ||
AJ238214.2 | - | 3 - 4 | 234 | 108 | ||
AJ292465.1 | - | 3 - 4 | 214 | 108 | ||
AJ292466.1 | - | 3 - 4 | 214 | 108 | ||
BC064602.1 | - | 3 - 4 | 266 | 108 | ||
BC142986.1 | - | 3 - 4 | 279 | 108 | ||
BI914338.1 | - | 276 | 108 | |||
DA792365.1 | - | 3 - 4 | 466 | 108 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000459.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017028373.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
