Assay Details
| Gene Symbol: TMEFF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003692.4 | NP_003683.2 | 1 - 2 | 503 | 126 | |
| GenBank mRNA | BC035056.1 | - | 1 - 2 | 305 | 5262325 | 126 |
| DN602026.1 | - | 1 - 2 | 503 | 126 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09002903.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005622.hg.1 | TC09002903.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005622.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005622.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005626.hg.1 | TC09002903.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005626.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005626.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005625.hg.1 | TC09002903.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005625.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09005625.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184347.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184347.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184347.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184347.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184347.hg.1 | TC0900012154.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012154.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184345.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184345.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184345.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184345.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184345.hg.1 | TC0900012154.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900184344.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184344.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184344.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184344.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184344.hg.1 | TC0900012154.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900184343.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184343.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184343.hg.1 | ENST00000374879.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184343.hg.1 | NM_003692.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184343.hg.1 | TC0900012154.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | ENST00000502978.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012155.hg.1 | NM_001198812.1 | Refseq |

TMEFF1
Genomic Map