Assay Details
| Gene Symbol: PHYHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014759.3 | NP_055574.3 | 1 - 2 | 541 | 70 | |
| XM_006716416.1 | XP_006716479.1 | 1 - 2 | 336 | 70 | ||
| GenBank mRNA | AK098743.1 | - | 2 - 3 | 286 | 70 | |
| AK294372.1 | - | 1 - 2 | 272 | 70 | ||
| BC034034.1 | - | 1 - 2 | 294 | 4793002 | 70 | |
| BC035940.1 | - | 1 - 2 | 294 | 4793228 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001036.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294372.1,BC034034.1,BC035940.1,NM_014759.3,XM_006716416.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | TC08001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | ENST00000321613.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | ENST00000454243.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | ENST00000456132.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013242.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013245.hg.1 | TC08001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013245.hg.1 | ENST00000454243.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013245.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08013245.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166819.hg.1 | TC0800009833.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800166817.hg.1 | TC0800009833.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000321613.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000454243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000518274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000522869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | ENST00000523252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_001099335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009833.hg.1 | NM_014759.3 | Refseq |

PHYHIP
Genomic Map