Assay Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278945.1 | NP_001265874.1 | 1 - 2 | 381 | 98 | |
| XM_006716368.2 | XP_006716431.1 | 1 - 2 | 363 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278945.1, XM_006716368.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
mitotic nuclear division
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of stress-activated MAPK cascade
cellular response to UV
negative regulation of inflammatory response
protein serine/threonine kinase activity
protein binding
ATP binding
protein phosphorylation
mitotic nuclear division
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of stress-activated MAPK cascade
cellular response to UV
negative regulation of inflammatory response
protein serine/threonine kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167809.hg.1 | TC0800009970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009970.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167810.hg.1 | TC0800009970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000301905.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000521226.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000522944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | ENST00000524266.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | NM_001278945.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | NM_018492.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800167808.hg.1 | TC0800009970.hg.1 | Clariom D TC |

PBK
Genomic Map