| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08013941.hg.1 |
TC08001086.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08013941.hg.1 |
ENST00000301905.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08013941.hg.1 |
ENST00000522944.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08013941.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
ENST00000524266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009970.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000301905.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000521226.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
ENST00000522944.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
NM_001278945.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
NM_018492.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800167802.hg.1 |
TC0800009970.hg.1 |
Clariom D TC |