Assay Details
| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114123.2 | NP_001107595.1 | 1 - 2 | 183 | 102 | |
| NM_001257168.1 | NP_001244097.1 | 1 - 2 | 183 | 102 | ||
| GenBank mRNA | AF000672.1 | - | 1 - 2 | 176 | 102 | |
| CB852603.1 | - | 1 - 2 | 190 | 102 | ||
| M25269.1 | - | 1 - 2 | 176 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000999.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF000672.1,M25269.1,NM_001114123.2,NM_001257168.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
response to light stimulus
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cellular response to testosterone stimulus
cellular response to gamma radiation
response to fibroblast growth factor
positive regulation of neuron death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription from RNA polymerase II promoter
response to light stimulus
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cellular response to testosterone stimulus
cellular response to gamma radiation
response to fibroblast growth factor
positive regulation of neuron death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013278.hg.1 | TC0X000999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013278.hg.1 | ENST00000376983.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013278.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013278.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000247161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009581.hg.1 | NM_005229.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000343894.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000376983.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | ENST00000468956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | NM_001114123.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | NM_001257168.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165295.hg.1 | TC0X00009581.hg.1 | Clariom D TC |

ELK1
Genomic Map