Assay Details
| Gene Symbol: OC90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080399.2 | NP_001073868.2 | 8 - 9 | 717 | 81 | |
| GenBank mRNA | AK297911.1 | - | 9 - 10 | 717 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08002620.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08021404.hg.1 | TC08002620.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021404.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021404.hg.1 | ENST00000443356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021404.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021402.hg.1 | TC08002620.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021402.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021402.hg.1 | ENST00000443356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021402.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021401.hg.1 | TC08002620.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021401.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021401.hg.1 | ENST00000443356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021401.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188158.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188158.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188158.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188158.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188158.hg.1 | TC0800012467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012467.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188159.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188159.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188159.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188159.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188159.hg.1 | TC0800012467.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800188161.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188161.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188161.hg.1 | ENST00000254627.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188161.hg.1 | NM_001080399.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188161.hg.1 | TC0800012467.hg.1 | Clariom D TC |

OC90
Genomic Map