Assay Details
| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145095.1 | NP_001138567.1 | 12 - 13 | 1239 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08002621.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08021419.hg.1 | TC08002621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021419.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021419.hg.1 | ENST00000434736.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021419.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021420.hg.1 | TC08002621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021420.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021420.hg.1 | ENST00000434736.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08021420.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012468.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188179.hg.1 | TC0800012468.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000414222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000434736.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | ENST00000473291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | NM_001145095.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800188178.hg.1 | TC0800012468.hg.1 | Clariom D TC |

HHLA1
Genomic Map